Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152345.4 | NP_689558.4 | 6, 7 | 876 | ||
XM_005257920.4 | XP_005257977.1 | 6, 7 | 809 | |||
XM_011524319.2 | XP_011522621.1 | 6, 7 | 1046 | |||
XM_011524320.2 | XP_011522622.1 | 6, 7 | 808 | |||
XM_011524321.2 | XP_011522623.1 | 6, 7 | 808 | |||
XM_017024174.1 | XP_016879663.1 | 6, 7 | 743 | |||
XM_017024175.1 | XP_016879664.1 | 6, 7 | 745 | |||
XM_017024176.1 | XP_016879665.1 | 6, 7 | 675 | |||
GenBank mRNA | AK092673.1 | - | 6, 7 | 693 | ||
AK097716.1 | - | 5, 6 | 712 | |||
BC032554.1 | - | 6, 7 | 923 | 5557374 | ||
BC062597.1 | - | 5, 6 | 726 |