Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC75A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027159.1 | 1 | 322 | |||
NR_027160.1 | 1 | 322 | ||||
NR_027161.1 | 1 | 322 | ||||
NR_027162.1 | 1 | 322 | ||||
NR_027171.1 | 1 | 322 | ||||
NR_027667.1 | 1 | 322 | ||||
NR_045024.1 | 1 | 322 | ||||
NR_045027.1 | 1 | 322 | ||||
NR_045028.1 | 1 | 322 | ||||
GenBank mRNA | AK307982.1 | - | 1 | 272 | ||
BC027986.1 | - | 1 | 272 | 4940349 | ||
BC040159.1 | - | 1 | 251 | 5539041 | ||
BP377665.1 | - | 1 | 257 | |||
BQ954204.1 | - | 1 | 246 | |||
BU165462.1 | - | 1 | 249 | |||
CF126952.1 | - | 1 | 238 | |||
DA494627.1 | - | 1 | 273 | |||
DB258221.1 | - | 1 | 273 |