Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282417.1 | NP_001269346.1 | 8, 9 | 981 | ||
NM_152351.4 | NP_689564.3 | 9, 10 | 1031 | |||
XM_011523653.2 | XP_011521955.1 | 8, 9 | 1111 | |||
XM_017024191.1 | XP_016879680.1 | 9, 10 | 1188 | |||
GenBank mRNA | AK057946.1 | - | 9, 10 | 1032 | ||
BC039868.1 | - | 8, 9 | 998 | 4157703 |