Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCERG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040006.1 | NP_001035095.1 | 11 | 1812 | ||
NM_006706.3 | NP_006697.2 | 12 | 1875 | |||
XM_005268365.2 | XP_005268422.1 | 12 | 2061 | |||
XM_017008977.1 | XP_016864466.1 | 11 | 1998 | |||
XM_017008978.1 | XP_016864467.1 | 12 | 2061 | |||
XM_017008979.1 | XP_016864468.1 | 12 | 1874 | |||
XM_017008980.1 | XP_016864469.1 | 12 | 2061 | |||
XM_017008981.1 | XP_016864470.1 | 10 | 1467 | |||
XM_017008982.1 | XP_016864471.1 | 11 | 1792 | |||
XM_017008983.1 | XP_016864472.1 | 11 | 1792 | |||
XM_017008984.1 | XP_016864473.1 | 12 | 2061 | |||
XM_017008985.1 | XP_016864474.1 | 12 | 2061 | |||
GenBank mRNA | AB209910.1 | - | 11 | 1786 | ||
AF017789.1 | - | 12 | 1835 | |||
AK126455.1 | - | 10 | 1715 | |||
AK304319.1 | - | 11 | 1811 | |||
BC014151.2 | - | 11 | 1806 | |||
BC111727.1 | - | 11 | 1790 | 40028766 |