Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291488.1 | NP_001278417.1 | 5 | 1170 | ||
NM_001291489.1 | NP_001278418.1 | 4 | 1240 | |||
NM_001291490.1 | NP_001278419.1 | 5 | 1304 | |||
NM_001291491.1 | NP_001278420.1 | 3 | 1022 | |||
NM_152354.5 | NP_689567.4 | 4 | 1149 | |||
GenBank mRNA | AK055309.1 | - | 1 | 758 | ||
AL833447.1 | - | 4 | 1127 | |||
BC069384.1 | - | 1 | 668 | 7262358 | ||
BC074824.2 | - | 1 | 631 | |||
BC074875.2 | - | 1 | 631 | 30915235 | ||
BC117300.1 | - | 3 | 1113 | 40125851 | ||
BC117302.1 | - | 3 | 1113 | 40125853 | ||
BC143984.1 | - | 4 | 1134 |