Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152355.2 | NP_689568.2 | 4 | 718 | ||
GenBank mRNA | AK095956.1 | - | 5 | 691 | ||
BC144217.1 | - | 3 | 514 |