Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152383.4 | NP_689596.4 | 20, 21 | 2780 | ||
NR_046476.1 | 20, 21 | 2707 | ||||
NR_046477.1 | 19 | 3280 | ||||
GenBank mRNA | AK090419.1 | - | 4 | 3833 | ||
AL834174.1 | - | 23 | 3986 | |||
BC026166.1 | - | 20, 21 | 2611 | 4822064 | ||
BC030113.2 | - | 3, 4 | 359 | |||
BC036113.1 | - | 20, 21 | 2606 | 5297794 | ||
BX648325.1 | - | 20 | 3171 |