Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM178A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152390.2 | NP_689603.2 | 1, 2 | 488 | ||
XM_005264144.1 | XP_005264201.1 | 1, 2 | 466 | |||
XM_017003369.1 | XP_016858858.1 | 1, 2 | 466 | |||
GenBank mRNA | AK075440.1 | - | 1, 2 | 464 | ||
AY358773.1 | - | 2, 3 | 279 | |||
BC029530.1 | - | 1, 2 | 433 | 5286077 | ||
DN993805.1 | - | 1, 2 | 494 |