Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014446.2 | NP_001014446.1 | 3 | 382 | ||
NM_001286773.1 | NP_001273702.1 | 3 | 405 | |||
NM_152398.3 | NP_689611.1 | 3 | 382 | |||
GenBank mRNA | AK298305.1 | - | 3 | 351 | ||
AY472117.1 | - | 3 | 384 | |||
BC032808.1 | - | 3 | 353 | 4454708 | ||
BG748993.1 | - | 3 | 289 | |||
BI546814.1 | - | 3 | 387 | |||
BM782144.1 | - | 3 | 352 | |||
CR749310.1 | - | 3 | 327 | |||
W68799.1 | - | 3 | 334 |