Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM69B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152421.3 | NP_689634.2 | 2, 3 | 291 | ||
XM_006716954.3 | XP_006717017.1 | 2, 3 | 296 | |||
XM_011518238.2 | XP_011516540.1 | 3, 4 | 422 | |||
GenBank mRNA | AJ420457.1 | - | 1, 2 | 131 | ||
BC032097.2 | - | 2, 3 | 285 | 3161575 | ||
BG700129.1 | - | 2, 3 | 296 |