Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152424.3 | NP_689637.3 | 2 | 363 | ||
GenBank mRNA | AK097146.1 | - | 2 | 363 | ||
DA939151.1 | - | 2 | 363 | |||
EF186024.1 | - | 2 | 224 |