Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282613.1 | NP_001269542.1 | 3 | 568 | ||
NM_001282614.1 | NP_001269543.1 | 5 | 830 | |||
NM_032735.2 | NP_116124.2 | 5 | 830 | |||
NM_152439.3 | NP_689652.2 | 2 | 330 | |||
XM_006719256.3 | XP_006719319.1 | 4 | 618 | |||
XM_011537961.1 | XP_011536263.1 | 3 | 438 | |||
XM_011537962.2 | XP_011536264.1 | 2 | 201 | |||
XM_011537963.1 | XP_011536265.1 | 2 | 209 | |||
GenBank mRNA | AF440758.1 | - | 5 | 775 | ||
AK096459.1 | - | 5 | 831 | |||
AY515706.1 | - | 4 | 603 | |||
BC151138.1 | - | 3 | 437 |