Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152339.3 | NP_689552.2 | 2 | 380 | ||
XM_005256279.4 | XP_005256336.1 | 2 | 797 | |||
GenBank mRNA | AK127372.1 | - | 1 | 184 | ||
BC027606.1 | - | 2 | 377 | |||
BC039608.1 | - | 2 | 358 | 5248678 | ||
BC053588.1 | - | 2 | 362 | 5245683 | ||
BI917028.1 | - | 2 | 362 |