Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM213B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195736.1 | NP_001182665.1 | 7 | 1055 | ||
NM_001195737.1 | NP_001182666.1 | 7 | 989 | |||
NM_001195738.1 | NP_001182667.1 | 7 | 969 | |||
NM_001195740.1 | NP_001182669.1 | 6 | 893 | |||
NM_001195741.1 | NP_001182670.1 | 6 | 890 | |||
NM_152371.3 | NP_689584.2 | 7 | 1001 | |||
NR_036637.1 | 7 | 934 | ||||
NR_036638.1 | 7 | 902 | ||||
XM_006710354.3 | XP_006710417.1 | 7 | 989 | |||
XM_011540664.1 | XP_011538966.1 | 7 | 1045 | |||
XM_011540665.1 | XP_011538967.1 | 7 | 1013 | |||
XM_011540666.1 | XP_011538968.1 | 7 | 993 | |||
GenBank mRNA | AF425266.1 | - | 7 | 944 | ||
AK057027.1 | - | 7 | 986 | |||
AK075273.1 | - | 7 | 921 | |||
AK094901.1 | - | 6 | 803 | |||
AK291908.1 | - | 7 | 927 | |||
AK298926.1 | - | 6 | 844 | |||
AK303504.1 | - | 7 | 961 | |||
AK308034.1 | - | 7 | 911 | |||
AK316243.1 | - | 6 | 803 | |||
BC022547.1 | - | 7 | 941 | 4812314 | ||
CR749706.1 | - | 2 | 733 |