Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152377.2 | NP_689590.1 | 3 | 292 | ||
XM_017000307.1 | XP_016855796.1 | 2 | 102 | |||
XM_017000308.1 | XP_016855797.1 | 3 | 283 | |||
GenBank mRNA | AK124828.1 | - | 3 | 277 | ||
BC027878.1 | - | 3 | 276 | 5207590 | ||
CD707889.1 | - | 3 | 328 |