Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199144.1 | NP_001186073.1 | 7 | 9136 | ||
NM_152381.5 | NP_689594.4 | 9 | 9538 | |||
XM_017003309.1 | XP_016858798.1 | 7 | 9033 | |||
GenBank mRNA | AK056582.1 | - | 1 | 766 | ||
AL831970.2 | - | 1 | 1740 | |||
AL831985.1 | - | 1 | 1400 | |||
AL832011.1 | - | 1 | 2024 | |||
AL832336.1 | - | 1 | 2065 | |||
AL832360.1 | - | 1 | 1645 | |||
AL832375.1 | - | 1 | 1075 | |||
AL832382.1 | - | 1 | 1592 | |||
AL832452.2 | - | 2 | 2113 | |||
AL833291.2 | - | 2 | 3356 | |||
AY820969.1 | - | 7 | 9005 | |||
BC070143.1 | - | 1 | 690 | |||
BC107799.1 | - | 1 | 608 | |||
BX647448.1 | - | 1 | 3088 | |||
BX647645.1 | - | 1 | 1759 | |||
BX647844.1 | - | 1 | 2939 | |||
BX647845.1 | - | 1 | 1697 | |||
BX647848.1 | - | 1 | 3577 | |||
EF119711.1 | - | 7 | 9136 |