Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039693.2 | NP_001034782.1 | 7 | 913 | ||
NR_004862.1 | 7 | 894 | ||||
NR_109905.1 | 4 | 642 | ||||
NR_109906.1 | 6 | 806 | ||||
NR_109907.1 | 6 | 758 | ||||
XM_017003310.1 | XP_016858799.1 | 5 | 541 | |||
GenBank mRNA | AK095272.1 | - | 6 | 522 | ||
AK293064.1 | - | 6 | 573 | |||
AK298977.1 | - | 7 | 657 | |||
BC025758.1 | - | 6 | 553 | |||
BC063502.1 | - | 3 | 182 | |||
BC132835.1 | - | 7 | 677 | 40146857 | ||
BC136837.1 | - | 7 | 677 | 9020828 | ||
BC144420.1 | - | 7 | 658 | |||
BX647677.1 | - | 7 | 896 | |||
CN261486.1 | - | 7 | 626 | |||
HY182974.1 | - | 4 | 417 |