Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152385.2 | NP_689598.2 | 3 | 484 | ||
XM_005264136.2 | XP_005264193.1 | 2 | 352 | |||
XM_011532519.2 | XP_011530821.1 | 3 | 3377 | |||
XM_011532520.2 | XP_011530822.1 | 3 | 655 | |||
XM_011532521.2 | XP_011530823.1 | 4 | 811 | |||
XM_011532522.2 | XP_011530824.1 | 4 | 798 | |||
XM_011532523.2 | XP_011530825.1 | 3 | 887 | |||
XM_011532524.1 | XP_011530826.1 | 2 | 269 | |||
XM_011532525.2 | XP_011530827.1 | 2 | 335 | |||
XM_011532527.2 | XP_011530829.1 | 3 | 3377 | |||
XM_011532528.1 | XP_011530830.1 | 2 | 277 | |||
XM_011532534.1 | XP_011530836.1 | 2 | 352 | |||
XM_011532535.2 | XP_011530837.1 | 1 | 50 | |||
XM_017003333.1 | XP_016858822.1 | 3 | 3377 | |||
XM_017003334.1 | XP_016858823.1 | 3 | 655 | |||
XM_017003339.1 | XP_016858828.1 | 3 | 3377 | |||
XM_017003340.1 | XP_016858829.1 | 3 | 3377 | |||
XM_017003341.1 | XP_016858830.1 | 2 | 496 | |||
XM_017003343.1 | XP_016858832.1 | 2 | 277 | |||
XM_017003346.1 | XP_016858835.1 | 3 | 3377 | |||
XM_017003348.1 | XP_016858837.1 | 3 | 3377 | |||
GenBank mRNA | AK056000.1 | - | 2 | 259 | ||
AK315683.1 | - | 3 | 442 | |||
BC029502.1 | - | 3 | 484 | 5270720 | ||
DA713663.1 | - | 2 | 246 |