Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320833.1 | NP_001307762.1 | 4 | 655 | ||
NM_001320834.1 | NP_001307763.1 | 3 | 224 | |||
NM_152386.3 | NP_689599.2 | 3 | 512 | |||
XM_017003355.1 | XP_016858844.1 | 3 | 414 | |||
GenBank mRNA | AF542512.1 | - | 3 | 416 | ||
AK096323.1 | - | 3 | 226 | |||
AK314322.1 | - | 3 | 512 | |||
BC106904.1 | - | 3 | 407 | |||
BC106905.1 | - | 3 | 409 | |||
BC134342.1 | - | 3 | 416 | 40031702 |