Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317837.1 | NP_001304766.1 | 5 | 579 | ||
NM_001317838.1 | NP_001304767.1 | 5 | 576 | |||
NM_001317839.1 | NP_001304768.1 | 5 | 454 | |||
NM_001317841.1 | NP_001304770.1 | 4 | 869 | |||
NM_001317842.1 | NP_001304771.1 | 4 | 713 | |||
NM_152399.3 | NP_689612.3 | 5 | 735 | |||
XM_005262737.2 | XP_005262794.1 | 5 | 543 | |||
XM_017007725.1 | XP_016863214.1 | 5 | 449 | |||
XM_017007726.1 | XP_016863215.1 | 4 | 389 | |||
GenBank mRNA | AK055396.1 | - | 5 | 456 | ||
AK310583.1 | - | 4 | 588 | |||
AL552582.3 | - | 5 | 606 | |||
BC047657.1 | - | 5 | 735 | 5264721 | ||
BX359599.2 | - | 4 | 741 | |||
DA176165.1 | - | 5 | 540 |