Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250790.2 | XP_005250847.1 | 2, 3 | 320 | ||
GenBank mRNA | BC028166.1 | - | 2, 3 | 320 |