Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286657.1 | NP_001273586.1 | 2 | 400 | ||
NM_001286660.1 | NP_001273589.1 | 3 | 539 | |||
NM_152417.2 | NP_689630.1 | 3 | 539 | |||
XM_005251150.1 | XP_005251207.1 | 3 | 528 | |||
XM_011517454.1 | XP_011515756.1 | 3 | 552 | |||
XM_011517455.2 | XP_011515757.1 | 3 | 528 | |||
GenBank mRNA | AK056932.1 | - | 3 | 458 | ||
AK308247.1 | - | 3 | 441 | |||
BC020835.1 | - | 3 | 498 | 4806471 | ||
BC071636.1 | - | 3 | 499 | 5193338 |