Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARNMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152420.2 | NP_689633.1 | 1 | 278 | ||
NR_135282.1 | 1 | 278 | ||||
XM_011518223.2 | XP_011516525.1 | 1 | 304 | |||
GenBank mRNA | AK299691.1 | - | 1 | 121 | ||
BC034033.1 | - | 1 | 242 | 4278547 | ||
BI560441.1 | - | 1 | 283 | |||
BX647386.1 | - | 1 | 47 |