Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152345.4 | NP_689558.4 | 15 | 3075 | ||
XM_005257920.4 | XP_005257977.1 | 15 | 3041 | |||
XM_011524319.2 | XP_011522621.1 | 15 | 3278 | |||
XM_017024174.1 | XP_016879663.1 | 15 | 2975 | |||
XM_017024175.1 | XP_016879664.1 | 15 | 2977 | |||
XM_017024176.1 | XP_016879665.1 | 15 | 2907 | |||
GenBank mRNA | AK000425.1 | - | 3 | 1434 | ||
AK092673.1 | - | 15 | 2891 | |||
AK097716.1 | - | 15 | 2368 | |||
AK098421.1 | - | 6 | 1581 | |||
BC032554.1 | - | 16 | 2579 | 5557374 | ||
BC062597.1 | - | 14 | 2925 |