Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC43A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284498.1 | NP_001271427.1 | 11, 12 | 1580 | ||
NM_001284499.1 | NP_001271428.1 | 6, 7 | 1021 | |||
NM_001321364.1 | NP_001308293.1 | 11, 12 | 1378 | |||
NM_001321365.1 | NP_001308294.1 | 11, 12 | 1547 | |||
NM_152346.2 | NP_689559.1 | 10, 11 | 1366 | |||
XM_017024177.1 | XP_016879666.1 | 10, 11 | 1543 | |||
XM_017024178.1 | XP_016879667.1 | 10, 11 | 1576 | |||
XM_017024179.1 | XP_016879668.1 | 9, 10 | 1475 | |||
GenBank mRNA | AB120364.1 | - | 11, 12 | 1546 | ||
AK075068.1 | - | 8, 9 | 906 | |||
AK075173.1 | - | 8, 9 | 960 | |||
AK075196.1 | - | 4, 5 | 647 | |||
AK301127.1 | - | 6, 7 | 904 | |||
BC019562.1 | - | 4, 5 | 641 | |||
BC027923.1 | - | 10, 11 | 1316 | 5309751 |