Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK074111.1 | - | 9 | 4271 | ||
AK091136.1 | - | 11 | 3632 | |||
BC114519.1 | - | 9 | 2906 | 40036610 | ||
BC114568.1 | - | 10 | 2973 | 40036619 |