Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321449.1 | NP_001308378.1 | 5 | 553 | ||
NM_152358.2 | NP_689571.2 | 5 | 553 | |||
XM_011526436.2 | XP_011524738.1 | 5 | 493 | |||
XM_011526437.2 | XP_011524739.1 | 4 | 487 | |||
GenBank mRNA | AY358196.1 | - | 5 | 467 | ||
BC029535.1 | - | 5 | 434 | |||
DB523757.2 | - | 5 | 325 |