Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM178A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167959.1 | NP_001161431.1 | 4 | 1148 | ||
NM_152390.2 | NP_689603.2 | 4 | 1485 | |||
XM_005264144.1 | XP_005264201.1 | 3 | 1325 | |||
XM_005264145.1 | XP_005264202.1 | 2 | 1211 | |||
XM_017003370.1 | XP_016858859.1 | 4 | 1267 | |||
XM_017003371.1 | XP_016858860.1 | 4 | 1184 | |||
GenBank mRNA | AK075440.1 | - | 4 | 1461 | ||
AY358773.1 | - | 5 | 1091 | |||
BC029530.1 | - | 4 | 1430 | 5286077 |