Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152401.2 | NP_689614.2 | 4 | 345 | ||
XM_005265728.3 | XP_005265785.1 | 4 | 407 | |||
XM_011534374.2 | XP_011532676.1 | 4 | 324 | |||
XM_017007745.1 | XP_016863234.1 | 4 | 408 | |||
GenBank mRNA | BC034431.1 | - | 4 | 345 | 5166825 |