Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOT1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152413.2 | NP_689626.2 | 9 | 1205 | ||
XM_005273399.3 | XP_005273456.1 | 9 | 1574 | |||
XM_006716285.3 | XP_006716348.1 | 8 | 1485 | |||
XM_011544391.2 | XP_011542693.1 | 6 | 661 | |||
GenBank mRNA | BC029504.1 | - | 9 | 1206 | 5273324 |