Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWF19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152434.2 | NP_689647.2 | 13 | 2011 | ||
XM_017017239.1 | XP_016872728.1 | 12 | 1329 | |||
GenBank mRNA | AK056905.1 | - | 11 | 1663 | ||
AK126881.1 | - | 13 | 2002 | |||
AK293001.1 | - | 13 | 1946 | |||
BC056241.1 | - | 1 | 106 | |||
BC110439.1 | - | 10 | 1624 | 40001346 | ||
BC110440.1 | - | 10 | 1624 | 40001348 | ||
BC110441.1 | - | 10 | 1566 | 40001349 | ||
BC110442.1 | - | 10 | 1624 | 40001350 | ||
BC118669.1 | - | 10 | 1627 | 40012545 | ||
BX647832.1 | - | 13 | 1985 | |||
BX648704.1 | - | 13 | 1990 | |||
BX649022.1 | - | 13 | 1981 |