Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204298.1 | NP_001191227.1 | 5 | 3236 | ||
NM_152437.2 | NP_689650.1 | 5 | 3288 | |||
GenBank mRNA | AK023009.1 | - | 1 | 1581 | ||
AL834266.1 | - | 6 | 4089 | |||
BC051696.1 | - | 5 | 3218 | 5276234 | ||
BX647762.1 | - | 6 | 3208 |
Gene Symbol: C10orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB073653.1 | - | 2122 |