Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304949.1 | NP_001291878.1 | 4 | 535 | ||
NM_001304952.1 | NP_001291881.1 | 4 | 456 | |||
NM_001304953.1 | NP_001291882.1 | 4 | 754 | |||
NM_001304954.1 | NP_001291883.1 | 3 | 553 | |||
NM_152465.2 | NP_689678.1 | 3 | 489 | |||
XM_005257927.4 | XP_005257984.1 | 4 | 725 | |||
XM_006721723.3 | XP_006721786.1 | 4 | 731 | |||
XM_011524398.2 | XP_011522700.1 | 4 | 455 | |||
XM_011524399.2 | XP_011522701.1 | 3 | 370 | |||
XM_011524400.2 | XP_011522702.1 | 3 | 279 | |||
XM_011524401.1 | XP_011522703.1 | 2 | 280 | |||
XM_017024237.1 | XP_016879726.1 | 3 | 373 | |||
GenBank mRNA | AK301868.1 | - | 4 | 453 | ||
AK310299.1 | - | 3 | 547 | |||
AL137531.1 | - | 2 | 2754 | |||
BC029574.1 | - | 3 | 436 | 5267334 | ||
CD300282.1 | - | 4 | 529 | |||
CR984201.1 | - | 4 | 215 | |||
DB038438.1 | - | 4 | 480 |