Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM187B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152481.1 | NP_689694.1 | 1 | 162 | ||
GenBank mRNA | AK098526.1 | - | 1 | 162 | ||
BC039080.1 | - | 1 | 113 | |||
BC117457.1 | - | 1 | 166 | 40126008 | ||
BC117459.1 | - | 1 | 166 | 40126010 |