Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529914.2 | XP_011528216.1 | 5 | 733 | ||
XM_011529917.2 | XP_011528219.1 | 5 | 592 | |||
XM_011529918.1 | XP_011528220.1 | 5 | 733 | |||
XM_017028621.1 | XP_016884110.1 | 5 | 614 | |||
XM_017028622.1 | XP_016884111.1 | 5 | 533 | |||
GenBank mRNA | AK098703.1 | - | 5 | 558 | ||
BC030013.1 | - | 5 | 343 | 4823215 | ||
CR456341.1 | - | 4 | 320 | |||
HY002382.1 | - | 6 | 383 |