Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012384.4 | NP_036516.1 | 10 | 2498 | ||
XM_005260202.4 | XP_005260259.1 | 10 | 2709 | |||
XM_006723776.2 | XP_006723839.1 | 8 | 2150 | |||
XM_011528779.2 | XP_011527081.1 | 8 | 2214 | |||
GenBank mRNA | AB033095.1 | - | 10 | 2375 | ||
AL117510.1 | - | 1 | 878 | |||
AL133646.1 | - | 8 | 2222 |