Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095678.1 | - | 1 | 522 | ||
BC101449.1 | - | 1 | 521 | |||
BC101450.1 | - | 1 | 521 | |||
BC101451.1 | - | 1 | 521 | |||
BC113052.1 | - | 1 | 521 |
Gene Symbol: LINC01123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046111.1 | 2 | 1444 | |||
GenBank mRNA | BC101448.1 | - | 1 | 522 |