Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANSL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307976.1 | NP_001294905.1 | 5, 6 | 1806 | ||
NM_152519.3 | NP_689732.2 | 5, 6 | 1806 | |||
XM_005246328.4 | XP_005246385.1 | 5, 6 | 2088 | |||
XM_005246329.4 | XP_005246386.1 | 5, 6 | 1794 | |||
XM_005246330.4 | XP_005246387.1 | 5, 6 | 1675 | |||
XM_005246332.4 | XP_005246389.1 | 5, 6 | 2087 | |||
XM_011510706.2 | XP_011509008.1 | 5, 6 | 1713 | |||
XM_011510707.2 | XP_011509009.1 | 5, 6 | 1896 | |||
XM_011510710.2 | XP_011509012.1 | 4, 5 | 545 | |||
XM_017003430.1 | XP_016858919.1 | 5, 6 | 1832 | |||
GenBank mRNA | AK056911.1 | - | 5, 6 | 1693 | ||
AK074441.1 | - | 4, 5 | 576 | |||
BC032837.1 | - | 5, 6 | 1776 | 4829661 | ||
BC070141.1 | - | 4, 5 | 638 | |||
BC090854.1 | - | 4, 5 | 1144 | |||
BC126155.1 | - | 5, 6 | 1712 | 8991871 | ||
BC143914.1 | - | 5, 6 | 1712 | 9052432 |