Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2CR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168216.1 | NP_001161688.1 | 2 | 345 | ||
NM_001168217.1 | NP_001161689.1 | 2 | 345 | |||
NM_001168221.1 | NP_001161693.1 | 2 | 345 | |||
NM_152525.5 | NP_689738.3 | 2 | 345 | |||
GenBank mRNA | AK058080.1 | - | 2 | 317 | ||
AL833429.1 | - | 2 | 342 | |||
BC030659.2 | - | 2 | 338 | 4838801 | ||
BG701187.1 | - | 2 | 346 | |||
BG773299.1 | - | 2 | 341 | |||
BX648732.1 | - | 2 | 338 |