Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152533.2 | NP_689746.1 | 3 | 525 | ||
NR_130715.1 | 3 | 525 | ||||
XM_011512455.2 | XP_011510757.1 | 3 | 525 | |||
XM_011512457.2 | XP_011510759.1 | 3 | 525 | |||
XM_017005751.1 | XP_016861240.1 | 3 | 495 | |||
XM_017005752.1 | XP_016861241.1 | 3 | 357 | |||
XM_017005753.1 | XP_016861242.1 | 3 | 525 | |||
GenBank mRNA | AK097841.1 | - | 3 | 525 | ||
BC032025.1 | - | 3 | 475 | 5165128 | ||
DB055549.1 | - | 3 | 525 |