Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152447.3 | NP_689660.2 | 3 | 1511 | ||
XM_005267369.3 | XP_005267426.1 | 3 | 4388 | |||
XM_017021048.1 | XP_016876537.1 | 4 | 4327 | |||
XM_017021049.1 | XP_016876538.1 | 2 | 2213 | |||
XM_017021050.1 | XP_016876539.1 | 2 | 4212 | |||
GenBank mRNA | AK055365.1 | - | 3 | 831 | ||
AK096627.1 | - | 3 | 1484 | |||
AL832129.1 | - | 5 | 2745 | |||
BC043165.1 | - | 3 | 936 | 5286818 |