Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284417.1 | NP_001271346.1 | 3 | 1111 | ||
NM_001284418.1 | NP_001271347.1 | 3 | 1112 | |||
NM_001284419.1 | NP_001271348.1 | 5 | 1187 | |||
NM_001284420.1 | NP_001271349.1 | 2 | 821 | |||
NM_001284421.1 | NP_001271350.1 | 4 | 1137 | |||
NM_001284422.1 | NP_001271351.1 | 5 | 1237 | |||
NM_152449.3 | NP_689662.2 | 6 | 1302 | |||
XM_011521241.1 | XP_011519543.1 | 2 | 895 | |||
XM_011521244.1 | XP_011519546.1 | 3 | 1464 | |||
XM_017021926.1 | XP_016877415.1 | 4 | 1328 | |||
XM_017021927.1 | XP_016877416.1 | 3 | 1649 | |||
XM_017021928.1 | XP_016877417.1 | 2 | 1293 | |||
XM_017021929.1 | XP_016877418.1 | 4 | 1490 | |||
XM_017021930.1 | XP_016877419.1 | 4 | 1504 | |||
GenBank mRNA | AK057570.1 | - | 1 | 646 | ||
AK124852.1 | - | 4 | 1440 | |||
BC041097.1 | - | 6 | 1279 | 5240362 | ||
BC084545.1 | - | 3 | 1079 | 4590701 |