Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM81A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152450.2 | NP_689663.2 | 3 | 422 | ||
XM_005254166.2 | XP_005254223.1 | 3 | 335 | |||
XM_006720398.3 | XP_006720461.1 | 5 | 604 | |||
XM_006720399.2 | XP_006720462.1 | 3 | 395 | |||
XM_011521247.2 | XP_011519549.1 | 5 | 568 | |||
XM_011521248.2 | XP_011519550.1 | 4 | 449 | |||
XM_011521249.2 | XP_011519551.1 | 4 | 559 | |||
XM_011521250.2 | XP_011519552.1 | 4 | 412 | |||
XM_017021931.1 | XP_016877420.1 | 4 | 860 | |||
XM_017021932.1 | XP_016877421.1 | 3 | 450 | |||
GenBank mRNA | AK299220.1 | - | 3 | 394 | ||
AK307975.1 | - | 3 | 365 | |||
BC022537.1 | - | 3 | 422 | 4817144 |