Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152457.2 | NP_689670.1 | 4 | 408 | ||
GenBank mRNA | AK057633.1 | - | 4 | 373 | ||
AL701654.1 | - | 4 | 291 | |||
BC029899.1 | - | 4 | 361 | 5175734 |