Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281293.1 | NP_001268222.1 | 2 | 2747 | ||
NM_001281294.1 | NP_001268223.1 | 3 | 2639 | |||
NM_012482.4 | NP_036614.1 | 2 | 2743 | |||
XM_017000887.1 | XP_016856376.1 | 5 | 4128 | |||
XM_017000888.1 | XP_016856377.1 | 5 | 3086 | |||
GenBank mRNA | AF086540.1 | - | 1 | 168 | ||
AF125158.1 | - | 1 | 2639 | |||
AJ132591.1 | - | 1 | 2160 | |||
AJ132592.1 | - | 2 | 2723 | |||
AK021707.1 | - | 1 | 1046 | |||
AK022916.1 | - | 2 | 2674 | |||
AK094701.1 | - | 3 | 2574 | |||
AK297233.1 | - | 3 | 2597 | |||
BC051905.1 | - | 1 | 2632 | 6165292 | ||
BC060820.1 | - | 2 | 2723 | 30344180 |