Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159860.1 | NP_001153332.1 | 3 | 292 | ||
NM_001159861.1 | NP_001153333.1 | 3 | 297 | |||
NM_152478.2 | NP_689691.2 | 3 | 344 | |||
XM_005258558.1 | XP_005258615.1 | 1 | 130 | |||
XM_011526517.2 | XP_011524819.1 | 3 | 2128 | |||
XM_017026350.1 | XP_016881839.1 | 3 | 273 | |||
XM_017026351.1 | XP_016881840.1 | 3 | 447 | |||
XM_017026352.1 | XP_016881841.1 | 3 | 424 | |||
XM_017026353.1 | XP_016881842.1 | 3 | 2206 | |||
GenBank mRNA | AK055592.1 | - | 3 | 320 | ||
BC111772.1 | - | 2 | 226 | 40034375 | ||
BP247825.1 | - | 3 | 279 | |||
DA407197.1 | - | 3 | 292 | |||
DA785623.1 | - | 3 | 347 | |||
DA942855.1 | - | 3 | 297 |