Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152492.2 | NP_689705.2 | 1 | 184 | ||
XM_011540780.1 | XP_011539082.1 | 1 | 187 | |||
XM_017000402.1 | XP_016855891.1 | 1 | 187 | |||
XM_017000403.1 | XP_016855892.1 | 1 | 187 | |||
XM_017000404.1 | XP_016855893.1 | 1 | 187 | |||
XM_017000405.1 | XP_016855894.1 | 1 | 187 | |||
GenBank mRNA | AK057387.1 | - | 1 | 187 | ||
BC112108.1 | - | 1 | 185 | 8327576 | ||
BC112112.1 | - | 1 | 184 | 8327580 | ||
DB048639.1 | - | 1 | 142 |