Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152493.2 | NP_689706.2 | 9 | 2844 | ||
XM_005270507.2 | XP_005270564.1 | 8 | 2984 | |||
XM_005270508.4 | XP_005270565.1 | 9 | 3641 | |||
XM_017000415.1 | XP_016855904.1 | 8 | 3556 | |||
GenBank mRNA | AF318356.1 | - | 8 | 2694 | ||
AK098342.1 | - | 1 | 1083 | |||
BC071632.1 | - | 2 | 1656 | |||
BC103704.1 | - | 1 | 1277 |