Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143687.2 | NP_001137159.1 | 4 | 397 | ||
NM_152494.3 | NP_689707.2 | 5 | 472 | |||
GenBank mRNA | AK057347.1 | - | 5 | 454 | ||
AK301876.1 | - | 4 | 374 | |||
AK301902.1 | - | 5 | 551 | |||
BC064844.1 | - | 4 | 427 | 30378702 |