Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303477.1 | NP_001290406.1 | 4 | 449 | ||
NM_001303478.1 | NP_001290407.1 | 4 | 428 | |||
NM_152504.3 | NP_689717.2 | 3 | 358 | |||
XM_006723549.3 | XP_006723612.1 | 3 | 289 | |||
XM_011529177.2 | XP_011527479.1 | 3 | 345 | |||
XM_017027684.1 | XP_016883173.1 | 4 | 460 | |||
GenBank mRNA | AK057796.1 | - | 3 | 355 | ||
AK292708.1 | - | 3 | 344 | |||
BC035800.1 | - | 3 | 348 | 5734487 | ||
BU656896.1 | - | 4 | 402 | |||
CD694965.1 | - | 3 | 357 | |||
HY080554.1 | - | 4 | 421 |